This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01312548_m1
See other UOX GE Assays ›
Gene Symbol:
Uox
Gene Name:
urate oxidase
Gene Aliases:
AI663847
Chromosome Location:
Chr.3: 146597149 - 146631483 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01312548_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Uox
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_009474.5 NP_033500.1 1 - 2 87 65
GenBank mRNA AK050425.1 - 1 - 2 88 65
BC019771.1 - 1 - 2 69 5136328 65
M27695.1 - 1 - 2 42 65

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0300001494.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Uox

Gene Ontology Categories:

Function(s) Process(es)

purine nucleobase metabolic process
oxidation-reduction process
urate oxidase activity
oxidoreductase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0300012114.mm.1 NM_009474.5.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300012114.mm.1 NM_009474.5 Refseq
Affymetrix Mouse Clariom S Array PSR0300012114.mm.1 TC0300001494.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0300012116.mm.1 NM_009474.5.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300012116.mm.1 NM_009474.5 Refseq
Affymetrix Mouse Clariom S Array PSR0300012116.mm.1 TC0300001494.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header