This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01312511_g1
See other ADGRL4 GE Assays ›
Gene Symbol:
Adgrl4
Gene Name:
adhesion G protein-coupled receptor L4
Gene Aliases:
1110033N21Rik, ETL1, Eltd1, Etl
Chromosome Location:
Chr.3: 151437882 - 151545081 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01312511_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Adgrl4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_133222.3 NP_573485.2 4 - 5 574 68
GenBank mRNA AF385682.1 - 4 - 5 577 68
BC017134.1 - 4 - 5 512 4511572 68

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0300001521.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Adgrl4

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

signal transduction
cell surface receptor signaling pathway
G-protein coupled receptor signaling pathway
signal transducer activity
transmembrane signaling receptor activity
G-protein coupled receptor activity
calcium ion binding
protein dimerization activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0300012251.mm.1 NM_133222.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300012251.mm.1 NM_133222.3 Refseq
Affymetrix Mouse Clariom S Array PSR0300012251.mm.1 TC0300001521.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0300012250.mm.1 NM_133222.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300012250.mm.1 NM_133222.3 Refseq
Affymetrix Mouse Clariom S Array PSR0300012250.mm.1 TC0300001521.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0