This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01312477_m1
See other TRPM1 GE Assays ›
Gene Symbol:
Trpm1
Gene Name:
transient receptor potential cation channel, subfamily M, member 1
Gene Aliases:
4732499L03Rik, AI606771, Ltrpc1, Mlsn1
Chromosome Location:
Chr.7: 64153835 - 64269772 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01312477_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Trpm1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001039104.2 NP_001034193.2 12 - 13 1843 61
NM_018752.3 NP_061222.3 12 - 13 1736 61
GenBank mRNA AF047714.1 - 12 - 13 1605 61
AK044507.1 - 12 - 13 1767 61
AK148193.1 - 10 - 11 1685 61
AK148268.1 - 12 - 13 1590 61
AK148275.1 - 12 - 13 1646 61
AK149290.1 - 12 - 13 1765 61
AK189982.1 - 2 - 3 164 61
AK190619.1 - 2 - 3 273 61
AK192027.1 - 2 - 3 164 61
AK194126.1 - 2 - 3 164 61
AK196486.1 - 2 - 3 164 61
AK199874.1 - 2 - 3 164 61
AK202089.1 - 2 - 3 169 61
AK202317.1 - 2 - 3 164 61
AK205137.1 - 2 - 3 170 61
AY180104.2 - 12 - 13 1843 61

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0700000976.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Ion Channel Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Trpm1

Panther Classification:

Molecular Function -

ion channel transporter

Gene Ontology Categories:

Function(s) Process(es)

transport
ion transport
signal transduction
G-protein coupled glutamate receptor signaling pathway
visual perception
ion transmembrane transport
retinal rod cell development
response to stimulus
transmembrane transport
calcium ion transport into cytosol
calcium ion transmembrane transport
cellular response to light stimulus
cation transmembrane transport
ion channel activity
cation channel activity
calcium channel activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700008883.mm.1 NM_001039104.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700008883.mm.1 NM_001039104.2 Refseq
Affymetrix Mouse Clariom S Array PSR0700008883.mm.1 NM_018752.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700008883.mm.1 NM_018752.3 Refseq
Affymetrix Mouse Clariom S Array PSR0700008883.mm.1 TC0700000976.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0