This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01312047_m1
See other PDE10A GE Assays ›
Gene Symbol:
Pde10a
Gene Name:
phosphodiesterase 10A
Gene Aliases:
-
Chromosome Location:
Chr.17: 8525187 - 8986648 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01312047_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pde10a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001290707.1 NP_001277636.1 7 - 8 911 74
NM_011866.2 NP_035996.2 6 - 7 865 74
GenBank mRNA AF110507.1 - 6 - 7 736 74
AK039249.1 - 7 - 8 911 74
AK147690.1 - 5 - 6 782 74
AK162221.1 - 6 - 7 520 74
AK162804.1 - 4 - 5 406 74
AY360383.1 - 6 - 7 865 74
BC113201.1 - 6 - 7 731 40048238 74

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1700000092.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Pde10a

Panther Classification:

Molecular Function -

phosphodiesterase

Gene Ontology Categories:

Function(s) Process(es)

cAMP catabolic process
signal transduction
metabolic process
regulation of protein kinase A signaling
regulation of cAMP-mediated signaling
cGMP catabolic process
nucleotide binding
catalytic activity
cyclic-nucleotide phosphodiesterase activity
3',5'-cyclic-nucleotide phosphodiesterase activity
cGMP-stimulated cyclic-nucleotide phosphodiesterase activity
phosphoric diester hydrolase activity
drug binding
hydrolase activity
cAMP binding
cGMP binding
metal ion binding
3',5'-cyclic-GMP phosphodiesterase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1700000793.mm.1 NM_011866.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700000793.mm.1 NM_011866.2 Refseq
Affymetrix Mouse Clariom S Array PSR1700000793.mm.1 TC1700000092.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0