This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01311399_g1
See other CENPF GE Assays ›
Gene Symbol:
Cenpf
Gene Name:
centromere protein F
Gene Aliases:
6530404A22Rik, AI325968, CENF, Lek1, PRO1779, mitosin
Chromosome Location:
Chr.1: 189640606 - 189688105 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01311399_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cenpf
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001081363.2 NP_001074832.2 14 - 15 8119 79
GenBank mRNA AF194970.1 - 3 - 4 6404 79
AK078350.1 - 1 - 2 130 79
BC023896.1 - 2 - 3 871 79
DQ642022.1 - 13 - 14 7976 79

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0100003783.mm.1

Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

mitotic cell cycle
chromosome segregation
regulation of G2/M transition of mitotic cell cycle
protein transport
regulation of striated muscle tissue development
negative regulation of transcription, DNA-templated
metaphase plate congression
protein C-terminus binding
transcription factor binding
protein homodimerization activity
dynein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0100030893.mm.1 NM_001081363.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100030893.mm.1 NM_001081363.2 Refseq
Affymetrix Mouse Clariom S Array PSR0100030893.mm.1 TC0100003783.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100030894.mm.1 NM_001081363.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100030894.mm.1 NM_001081363.2 Refseq
Affymetrix Mouse Clariom S Array PSR0100030894.mm.1 TC0100003783.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0