Assay Details
Gene Symbol: 4933425B07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_046460.1 | 4 - 5 | 421 | 66 | ||
GenBank mRNA | AK016907.1 | - | 4 - 5 | 422 | 66 | |
BY716586.1 | - | 4 - 5 | 422 | 66 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | RNA upstream of Bmp4 expressed in inner ear | Rubie | -27.0kb | Sense | Chr. 14 - 46568331 - 46575851 on NCBI Build GRCm38 | |
Downstream | cyclin-dependent kinase inhibitor 3 | Cdkn3 |
2410006H10Rik, KAP |
155.2kb | Anti-sense | Chr. 14 - 46760464 - 46771529 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC1500001464.rn
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2259456 | AK016907.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 694069 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 327534 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 360612 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 660734 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 802627 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 856591 | AK032445.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 410351 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 864434 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 845644 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 859602 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 868759 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 128645 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08100299 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08100273 | - | - |
Whole Mouse Genome G4122A | A_51_P153949 | AK016907.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P134196 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11652190 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29577326 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29577275 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29577285 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29577303 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29577485 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29577470 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29577295 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29577288 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29577335 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29577311 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29577312 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29577328 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29577293 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29577327 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29577309 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29577433 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11774645 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29577281 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11742279 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29577299 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29577347 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29577278 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29577337 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29577479 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11774639 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11420519 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11652189 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29577276 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11197788 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29577487 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29577357 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29577441 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29577493 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29577323 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11476051 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29577210 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29577297 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29577450 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29577305 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29577435 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11774652 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29577306 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29577301 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29577211 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29577209 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29577495 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29577469 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29577355 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29577340 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29577452 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11510607 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29577298 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29577489 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29577457 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29577307 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11774643 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29577345 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29577473 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29577290 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29577291 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29577449 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29577476 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29577351 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29577491 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29577314 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29577284 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29577444 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29577483 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29577331 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11159266 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29577333 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29577453 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29577436 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29577341 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29577458 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29577330 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29577352 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29577354 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11748450 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29577475 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29577454 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29577325 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29577322 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29577446 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29577439 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29577342 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix HT MG-430 PM Array Plate | 1453883_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1453883_at | - | - |
Mouse Genome 430 2.0 Array | 1453883_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1400014485.mm.1 | NR_046460.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1400014485.mm.1 | TC1400001917.mm.1 | MTA TC |
Mouse Expression Set 430 | 1453883_at | - | - |
Mouse Expression Set 430 | 1453883_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1453883_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1453883_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 452076 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 105670372 | - | - |
