This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01311154_m1
See other CEP170 GE Assays ›
Gene Symbol:
Cep170
Gene Name:
centrosomal protein 170
Gene Aliases:
4933426L22Rik, A330004A13Rik, AI195353
Chromosome Location:
Chr.1: 176733653 - 176814173 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01311154_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cep170
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001099637.2 NP_001093107.2 7 - 8 799 77

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0100003620.mm.1

Set Membership:

Amplicon length between 71 and 85 Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

scaffold/adaptor protein

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0100029749.mm.1 NM_001099637.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100029749.mm.1 NM_001099637.2 Refseq
Affymetrix Mouse Clariom S Array PSR0100029749.mm.1 TC0100003620.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100029751.mm.1 NM_001099637.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100029751.mm.1 NM_001099637.2 Refseq
Affymetrix Mouse Clariom S Array PSR0100029751.mm.1 TC0100003620.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0