This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01310569_m1
See other RB1 GE Assays ›
Gene Symbol:
Rb1
Gene Name:
retinoblastoma 1
Gene Aliases:
Rb, Rb-1, pRb
Chromosome Location:
Chr.14: 73194453 - 73325887 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01310569_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rb1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_009029.2 NP_033055.2 5 - 6 668 150
GenBank mRNA AK011246.1 - 5 - 6 668 150
AK141028.1 - 5 - 6 700 150
BC096525.1 - 5 - 6 668 6832808 150
DQ400415.1 - 5 - 6 518 150
M26391.1 - 5 - 6 616 150

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Rb1

Panther Classification:

Molecular Function -

chromatin/chromatin-binding, or -regulatory protein

Gene Ontology Categories:

Function(s) Process(es)

G1/S transition of mitotic cell cycle
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
negative regulation of protein kinase activity
cell cycle
cell cycle arrest
regulation of mitotic cell cycle
negative regulation of cell proliferation
chromatin modification
neuron differentiation
sister chromatid biorientation
neuron projection development
maintenance of mitotic sister chromatid cohesion
glial cell apoptotic process
skeletal muscle cell differentiation
neuron maturation
enucleate erythrocyte differentiation
regulation of lipid kinase activity
myoblast differentiation
positive regulation of macrophage differentiation
negative regulation of cell cycle
positive regulation of mitotic metaphase/anaphase transition
negative regulation of smoothened signaling pathway
negative regulation of transcription, DNA-templated
negative regulation of mitotic cell cycle
positive regulation of transcription from RNA polymerase II promoter
digestive tract development
cell morphogenesis involved in neuron differentiation
negative regulation of epithelial cell proliferation
striated muscle cell differentiation
cell division
neuron apoptotic process
regulation of cell cycle
protein localization to chromosome, centromeric region
cellular response to xenobiotic stimulus
regulation of cohesin loading
negative regulation of transcription involved in G1/S transition of mitotic cell cycle
hepatocyte apoptotic process
negative regulation of hepatocyte apoptotic process
negative regulation of G1/S transition of mitotic cell cycle
core promoter binding
RNA polymerase II activating transcription factor binding
DNA binding
protein binding
transcription factor binding
enzyme binding
kinase binding
ubiquitin protein ligase binding
identical protein binding
phosphoprotein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0