Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1400017070.mm.1 |
NM_001204340.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400017070.mm.1 |
NM_001204340.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400017070.mm.1 |
NM_172810.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400017070.mm.1 |
NM_172810.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400017070.mm.1 |
TC1400002206.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400017069.mm.1 |
NM_001204340.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400017069.mm.1 |
NM_001204340.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400017069.mm.1 |
NM_172810.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400017069.mm.1 |
NM_172810.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400017069.mm.1 |
TC1400002206.mm.1 |
MTA TC |