This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01310395_m1
See other SGMS2 GE Assays ›
Gene Symbol:
Sgms2
Gene Name:
sphingomyelin synthase 2
Gene Aliases:
4933405A16Rik, 5133401H06Rik, AI854299
Chromosome Location:
Chr.3: 131318985 - 131491192 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01310395_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Sgms2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_028943.5 NP_083219.2 4 - 5 1302 70
GenBank mRNA AK016659.1 - 4 - 5 1316 70
BC117782.2 - 4 - 5 1306 40092081 70

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0300002959.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Sgms2

Gene Ontology Categories:

Function(s) Process(es)

lipid metabolic process
sphingolipid metabolic process
sphingomyelin biosynthetic process
phosphorylation
kinase activity
transferase activity
sphingomyelin synthase activity
ceramide cholinephosphotransferase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0300023395.mm.1 NM_028943.5.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300023395.mm.1 NM_028943.5 Refseq
Affymetrix Mouse Clariom S Array PSR0300023395.mm.1 TC0300002959.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56ffd697dc-88d4p:80/100.66.130.94:80.
git-commit: 082a6d415be982b1f92c35e55dd71609d4576d3e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0