This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01309860_g1
See other RGS11 GE Assays ›
Gene Symbol:
Rgs11
Gene Name:
regulator of G-protein signaling 11
Gene Aliases:
C78048
Chromosome Location:
Chr.17: 26202962 - 26211324 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01309860_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rgs11
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001081069.1 NP_001074538.1 3 - 4 227 72
GenBank mRNA AF061934.1 - 2 - 3 132 72
AK039314.1 - 3 - 4 196 72
AK080707.1 - 3 - 4 317 72

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1700000407.mm.1

Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus 1700 Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

G-protein coupled receptor signaling pathway
negative regulation of signal transduction
intracellular signal transduction
termination of G-protein coupled receptor signaling pathway
positive regulation of GTPase activity
signal transducer activity
GTPase activator activity
protein binding
G-protein beta-subunit binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1700003442.mm.1 NM_001081069.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700003442.mm.1 NM_001081069.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700003442.mm.1 TC1700000407.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 439922 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56ffd697dc-88d4p:80/100.66.130.94:80.
git-commit: 082a6d415be982b1f92c35e55dd71609d4576d3e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0