This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01309588_g1
See other PFKM GE Assays ›
Gene Symbol:
Pfkm
Gene Name:
phosphofructokinase, muscle
Gene Aliases:
AI131669, ATP-PFK, PFK-A, PFK-M, Pfk-4, Pfk4, Pfka, Pfkx
Chromosome Location:
Chr.15: 98092589 - 98132447 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01309588_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pfkm
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001163487.1 NP_001156959.1 7 - 8 791 109
NM_001163488.1 NP_001156960.1 7 - 8 727 109
NM_021514.4 NP_067489.3 6 - 7 744 109
GenBank mRNA AF249894.1 - 6 - 7 684 109
AK002711.2 - 7 - 8 694 109
AK049773.1 - 7 - 8 699 109
BC005526.1 - 6 - 7 708 3495173 109
BC115774.1 - 4 - 5 499 109

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1500000969.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Pfkm

Panther Classification:

Molecular Function -

carbohydrate kinase kinase transferase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

fructose 6-phosphate metabolic process
glycolytic process
metabolic process
phosphorylation
fructose 1,6-bisphosphate metabolic process
positive regulation of insulin secretion
glucose homeostasis
muscle cell cellular homeostasis
carbohydrate phosphorylation
protein oligomerization
protein homotetramerization
glycolytic process through fructose-6-phosphate
nucleotide binding
catalytic activity
6-phosphofructokinase activity
protein binding
ATP binding
protein C-terminus binding
phosphofructokinase activity
AMP binding
kinase activity
transferase activity
kinase binding
carbohydrate binding
identical protein binding
protein homodimerization activity
metal ion binding
fructose binding
fructose-6-phosphate binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1500007613.mm.1 NM_001163487.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500007613.mm.1 NM_001163487.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500007613.mm.1 NM_001163488.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500007613.mm.1 NM_001163488.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500007613.mm.1 NM_021514.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500007613.mm.1 NM_021514.4 Refseq
Affymetrix Mouse Clariom S Array PSR1500007613.mm.1 TC1500000969.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0