This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01309170_m1
See other THOC6 GE Assays ›
Gene Symbol:
Thoc6
Gene Name:
THO complex 6 homolog (Drosophila)
Gene Aliases:
F830014G06Rik, Wdr58
Chromosome Location:
Chr.17: 23668618 - 23673903 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01309170_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Thoc6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001008425.1 NP_001008425.1 1 - 2 255 90
GenBank mRNA AK089746.1 - 1 - 2 328 90
BC085131.1 - 1 - 2 255 5715435 90
BC089551.1 - 2 - 3 255 30303151 90

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the BC085131.1, BC089551.1, NM_001008425.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Thoc6

Gene Ontology Categories:

Function(s) Process(es)

mRNA processing
mRNA export from nucleus
transport
apoptotic process
multicellular organism development
central nervous system development
RNA splicing
negative regulation of apoptotic process
viral mRNA export from host cell nucleus
mRNA transport
RNA binding
protein binding

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31092956 - -
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2169849 BC089551.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2169849 NM_001008425.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2169849 AK089746.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2169849 BC085131.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P631568 BC089551.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P631568 NM_001008425.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P631568 AK089746.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P631568 BC085131.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P631568 BC089551.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P631568 NM_001008425.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P631568 AK089746.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P631568 BC085131.1 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 288316 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 157241 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 485377 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 110491 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2169849 BC089551.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2169849 NM_001008425.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2169849 AK089746.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2169849 BC085131.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P631568 BC089551.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P631568 NM_001008425.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P631568 AK089746.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P631568 BC085131.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P631568 BC089551.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P631568 NM_001008425.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P631568 AK089746.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P631568 BC085131.1 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31092964 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31092960 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31092961 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31092958 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31092967 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03292144 - -
Whole Mouse Genome G4122A A_52_P631568 BC089551.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P631568 NM_001008425.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P631568 AK089746.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P631568 BC085131.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P631568 BC089551.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P631568 NM_001008425.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P631568 AK089746.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P631568 BC085131.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P113437 - -
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P111530 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13214697 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13214711 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13214726 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13214687 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31092956 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093028 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092874 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093010 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092886 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093004 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093008 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092955 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092877 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092870 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092884 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093020 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092960 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092869 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092868 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092967 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093005 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092863 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093019 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092996 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092976 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092867 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092979 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092883 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092872 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092866 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092961 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092876 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092862 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093022 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092989 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093014 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092985 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092964 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093017 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092958 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093029 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092865 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092990 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093011 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092978 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093003 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093016 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092998 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092969 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092880 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093001 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092881 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093024 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093006 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31092969 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31092964 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31092960 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31092955 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31092958 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31092961 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31092967 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10448306 - -
Affymetrix Mouse Gene 1.0 ST Array 10448295 - -
Affymetrix Mouse Gene 1.0 ST Array 10448293 - -
Affymetrix Mouse Gene 1.0 ST Array 10448296 - -
Affymetrix Mouse Gene 1.0 ST Array 10448305 - -
Affymetrix Mouse Gene 1.0 ST Array 10448299 - -
Affymetrix Mouse Gene 1.0 ST Array 10442322 - -
Affymetrix Mouse Gene 1.0 ST Array 10448303 - -
Affymetrix Mouse Gene 1.0 ST Array 10448298 - -
Affymetrix Mouse Gene 1.0 ST Array 10448300 - -
Affymetrix Mouse Gene 1.0 ST Array 10448301 - -
Affymetrix Mouse Gene 1.0 ST Array 10448297 - -
Affymetrix Mouse Gene 1.0 ST Array 10448302 - -
Affymetrix Mouse Gene 1.0 ST Array 10448304 - -
Affymetrix Mouse Gene 1.0 ST Array 10448294 - -
Affymetrix HT MG-430 PM Array Plate 1434827_PM_at - -
Affymetrix Mouse Gene 1.1 ST Array 10442321 - -
Affymetrix Mouse Gene 1.1 ST Array 10448292 - -
Mouse Genome 430 2.0 Array 1434827_PM_at - -
Mouse Genome 430 2.0 Array 1434827_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 386612_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 386612_PM_at - -
Affymetrix Mouse Clariom S Array PSR1700013959.mm.1 NM_001008425.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700013959.mm.1 NM_001008425.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700013959.mm.1 TC1700001597.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1700013953.mm.1 NM_001008425.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700013953.mm.1 NM_001008425.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700013953.mm.1 TC1700001597.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1700013958.mm.1 NM_001008425.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700013958.mm.1 NM_001008425.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700013958.mm.1 TC1700001597.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1700013955.mm.1 NM_001008425.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700013955.mm.1 NM_001008425.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700013955.mm.1 TC1700001597.mm.1 MTA TC
Mouse Expression Set 430 1434827_PM_at - -
Mouse Expression Set 430 1434827_at - -
Mouse Genome 430A 2.0 Array 1434827_PM_at - -
Mouse Genome 430A 2.0 Array 1434827_at - -
Affymetrix HT Mouse Genome 430A Array 1434827_PM_at - -
Affymetrix HT Mouse Genome 430A Array 1434827_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 693022 - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 100780600 - -
Illumina mouse-6 v1.0 expression beadchip 100780600 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_1249706 - -
Illumina Sentrix mouse-6.v1 bead array 100780600 - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 100780600 - -
MouseRef-8 v2 Expression BeadChip ILMN_2778428 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2778428 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0