Assay Details
Gene Symbol: Pifo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001200028.1 | NP_001186957.1 | 5 - 6 | 711 | 82 | |
NM_029604.3 | NP_083880.2 | 5 - 6 | 663 | 82 | ||
GenBank mRNA | AK006407.1 | - | 5 - 6 | 664 | 82 | |
AW492850.1 | - | 1 - 2 | 82 | |||
AW519725.1 | - | 4 - 5 | 375 | 82 | ||
BC049757.1 | - | 5 - 6 | 712 | 6773491 | 82 | |
HM237137.1 | - | 5 - 6 | 553 | 82 | ||
HM237138.1 | - | 5 - 6 | 711 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2054008 | BC049757.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2054008 | HM237138.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2054008 | NM_001200028.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2142990 | AK006407.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2142990 | HM237137.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2142990 | NM_029604.3 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 28918 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 2136 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 111087 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P135698 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P135664 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2142990 | AK006407.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2142990 | HM237137.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2142990 | NM_029604.3 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2054008 | BC049757.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2054008 | HM237138.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2054008 | NM_001200028.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22400467 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22400498 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22400483 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22400511 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22400507 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22400486 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22400475 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22400510 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22400489 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22400514 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22400470 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22400478 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22400524 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22400482 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22400481 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22400484 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22400487 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22400492 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22400505 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22400522 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22400501 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22400508 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22400494 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22400517 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22400468 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22400500 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22400497 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22400519 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22400516 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22400473 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22400515 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22400503 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22400512 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00719809 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P135664 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P135698 | - | - |
Whole Mouse Genome G4122A | A_51_P258901 | AK006407.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P258901 | AW519725.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P258901 | BC049757.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P258901 | HM237137.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P258901 | HM237138.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P135698 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P135664 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03056158 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03056159 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03056156 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03056148 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03056160 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03056147 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22400409 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22400393 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22400418 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22400438 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22400435 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22400387 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22400395 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22400461 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22400428 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22400448 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22400442 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22400452 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22400456 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22400440 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22400454 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22400390 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22400426 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22400429 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22400445 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22400355 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22400406 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22400458 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22400436 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22400430 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22400420 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22400422 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22400424 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22400397 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22400391 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22400416 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22400450 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22400386 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22400447 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22400457 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02359521 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22400430 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03126850 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22400452 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22400387 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22400442 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22400426 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22400386 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22400445 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02370334 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22400458 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22400420 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22400440 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03126855 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22400405 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02778542 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22400435 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22400403 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22400462 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22400406 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22400461 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03126853 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02894822 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22400460 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22400410 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22400436 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22400416 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22400424 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22400412 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22400414 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02698047 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22400443 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22400454 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22400438 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02371035 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22400450 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02709567 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22400422 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22400428 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22400418 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22400413 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22400459 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02424307 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22400456 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02719186 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10501002 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10501004 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10501006 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10501000 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10501003 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10501001 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10501005 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1453320_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10500999 | - | - |
Mouse Genome 430 2.0 Array | 1453320_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1453320_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0300020876.mm.1 | NM_001200028.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300020876.mm.1 | NM_001200028.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300020876.mm.1 | NM_029604.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300020876.mm.1 | NM_029604.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300020876.mm.1 | TC0300002682.mm.1 | MTA TC |
Mouse Expression Set 430 | 1453320_at | - | - |
Mouse Expression Set 430 | 1453320_PM_at | - | - |
Murine Genome U74v2 Set | 165845_PM_at | - | - |
Murine Genome U74v2 Set | 165845_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1453320_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1453320_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 390922 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2736521 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2736521 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2736521 | - | - |
