This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01307870_m1
See other PIFO GE Assays ›
Gene Symbol:
Pifo
Gene Name:
primary cilia formation
Gene Aliases:
1700027A23Rik
Chromosome Location:
Chr.3: 105996957 - 106014646 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01307870_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pifo
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001200028.1 NP_001186957.1 3 - 4 466 134
NM_029604.3 NP_083880.2 3 - 4 418 134
GenBank mRNA AK006407.1 - 3 - 4 419 134
AW519725.1 - 2 - 3 130 134
BC049757.1 - 3 - 4 467 6773491 134
HM237137.1 - 3 - 4 308 134
HM237138.1 - 3 - 4 466 134

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0300002682.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Pifo

Gene Ontology Categories:

Function(s) Process(es)

cell projection organization
regulation of cell projection organization
positive regulation of kinase activity
protein binding
Rab GTPase binding
kinesin binding
protein kinase binding
gamma-tubulin binding
beta-tubulin binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0300020878.mm.1 NM_001200028.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300020878.mm.1 NM_001200028.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300020878.mm.1 NM_029604.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300020878.mm.1 NM_029604.3 Refseq
Affymetrix Mouse Clariom S Array PSR0300020878.mm.1 TC0300002682.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0