This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01307342_m1
See other YLPM1 GE Assays ›
Gene Symbol:
Ylpm1
Gene Name:
YLP motif containing 1
Gene Aliases:
A930013E17Rik, ZAP, Zap3
Chromosome Location:
Chr.12: 84996285 - 85074533 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01307342_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ylpm1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_178363.3 NP_848140.3 11 - 12 5647 104
GenBank mRNA AB033168.1 - 11 - 12 3320 104
AK028731.1 - 1 - 2 82 104
AK044440.1 - 5 - 6 1964 104
AK162250.1 - 3 - 4 356 104

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1200000825.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ylpm1

Panther Classification:

Molecular Function -

nucleic acid metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
regulation of telomere maintenance
transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1200005965.mm.1 NM_178363.4 Refseq
Affymetrix Mouse Clariom S Array PSR1200005965.mm.1 TC1200000825.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0