This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01306817_m1
See other ZPBP2 GE Assays ›
Gene Symbol:
Zpbp2
Gene Name:
zona pellucida binding protein 2
Gene Aliases:
1700017D11Rik, 2610022C02Rik, ZPBPL
Chromosome Location:
Chr.11: 98551097 - 98558665 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01306817_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Zpbp2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001166494.1 NP_001159966.1 5 - 6 1002 93
NM_001166495.1 NP_001159967.1 6 - 7 791 93
NM_027061.2 NP_081337.2 6 - 7 882 93
NM_199419.3 NP_955451.2 5 - 6 663 93
GenBank mRNA BC050986.1 - 6 - 7 758 93
BC089476.1 - 5 - 6 663 6708062 93

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100001571.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Zpbp2

Gene Ontology Categories:

Function(s) Process(es)

acrosome assembly
binding of sperm to zona pellucida
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1100014568.mm.1 NM_001166494.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100014568.mm.1 NM_001166494.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100014568.mm.1 NM_001166495.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100014568.mm.1 NM_001166495.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100014568.mm.1 NM_027061.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100014568.mm.1 NM_027061.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100014568.mm.1 NM_199419.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100014568.mm.1 NM_199419.3 Refseq
Affymetrix Mouse Clariom S Array PSR1100014568.mm.1 TC1100001571.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100014561.mm.1 NM_001166494.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100014561.mm.1 NM_001166494.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100014561.mm.1 NM_001166495.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100014561.mm.1 NM_001166495.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100014561.mm.1 NM_027061.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100014561.mm.1 NM_027061.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100014561.mm.1 NM_199419.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100014561.mm.1 NM_199419.3 Refseq
Affymetrix Mouse Clariom S Array PSR1100014561.mm.1 TC1100001571.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100014557.mm.1 NM_001166494.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100014557.mm.1 NM_001166494.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100014557.mm.1 NM_001166495.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100014557.mm.1 NM_001166495.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100014557.mm.1 NM_027061.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100014557.mm.1 NM_027061.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100014557.mm.1 NM_199419.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100014557.mm.1 NM_199419.3 Refseq
Affymetrix Mouse Clariom S Array PSR1100014557.mm.1 TC1100001571.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100014560.mm.1 NM_001166494.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100014560.mm.1 NM_001166494.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100014560.mm.1 NM_001166495.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100014560.mm.1 NM_001166495.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100014560.mm.1 NM_027061.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100014560.mm.1 NM_027061.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100014560.mm.1 NM_199419.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100014560.mm.1 NM_199419.3 Refseq
Affymetrix Mouse Clariom S Array PSR1100014560.mm.1 TC1100001571.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100014572.mm.1 NM_001166494.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100014572.mm.1 NM_001166494.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100014572.mm.1 NM_001166495.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100014572.mm.1 NM_001166495.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100014572.mm.1 NM_027061.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100014572.mm.1 NM_027061.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100014572.mm.1 NM_199419.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100014572.mm.1 NM_199419.3 Refseq
Affymetrix Mouse Clariom S Array PSR1100014572.mm.1 TC1100001571.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100014564.mm.1 NM_001166494.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100014564.mm.1 NM_001166494.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100014564.mm.1 TC1100001571.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100014565.mm.1 NM_001166494.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100014565.mm.1 NM_001166494.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100014565.mm.1 NM_001166495.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100014565.mm.1 NM_001166495.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100014565.mm.1 NM_027061.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100014565.mm.1 NM_027061.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100014565.mm.1 NM_199419.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100014565.mm.1 NM_199419.3 Refseq
Affymetrix Mouse Clariom S Array PSR1100014565.mm.1 TC1100001571.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100014562.mm.1 NM_001166494.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100014562.mm.1 NM_001166494.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100014562.mm.1 NM_027061.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100014562.mm.1 NM_027061.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100014562.mm.1 NM_199419.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100014562.mm.1 NM_199419.3 Refseq
Affymetrix Mouse Clariom S Array PSR1100014562.mm.1 TC1100001571.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100014570.mm.1 NM_001166494.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100014570.mm.1 NM_001166494.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100014570.mm.1 NM_001166495.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100014570.mm.1 NM_001166495.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100014570.mm.1 NM_027061.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100014570.mm.1 NM_027061.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100014570.mm.1 TC1100001571.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100014563.mm.1 NM_001166494.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100014563.mm.1 NM_001166494.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100014563.mm.1 TC1100001571.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100014569.mm.1 NM_001166494.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100014569.mm.1 NM_001166494.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100014569.mm.1 NM_001166495.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100014569.mm.1 NM_001166495.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100014569.mm.1 NM_027061.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100014569.mm.1 NM_027061.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100014569.mm.1 NM_199419.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100014569.mm.1 NM_199419.3 Refseq
Affymetrix Mouse Clariom S Array PSR1100014569.mm.1 TC1100001571.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100014573.mm.1 NM_001166494.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100014573.mm.1 NM_001166494.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100014573.mm.1 NM_001166495.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100014573.mm.1 NM_001166495.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100014573.mm.1 NM_027061.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100014573.mm.1 NM_027061.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100014573.mm.1 NM_199419.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100014573.mm.1 NM_199419.3 Refseq
Affymetrix Mouse Clariom S Array PSR1100014573.mm.1 TC1100001571.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0