This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01306735_g1
See other GRB7 GE Assays ›
Gene Symbol:
Grb7
Gene Name:
growth factor receptor bound protein 7
Gene Aliases:
mKIAA4028
Chromosome Location:
Chr.11: 98446834 - 98455373 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01306735_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Grb7
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_010346.2 NP_034476.1 8 - 9 1349 55
GenBank mRNA AK057640.1 - 8 - 9 1342 55
AK145996.1 - 8 - 9 1165 55
AK168438.1 - 8 - 9 1133 55
AK220277.1 - 7 - 8 1240 55
BC003295.1 - 8 - 9 1092 3583896 55
M94450.1 - 8 - 9 1371 55

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100001570.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Grb7

Panther Classification:

Molecular Function -

scaffold/adaptor protein

Gene Ontology Categories:

Function(s) Process(es)

signal transduction
positive regulation of signal transduction
negative regulation of translation
positive regulation of cell migration
stress granule assembly
RNA binding
SH3/SH2 adaptor activity
protein binding
lipid binding
protein kinase binding
phosphatidylinositol binding
identical protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1100014543.mm.1 NM_010346.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100014543.mm.1 NM_010346.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100014543.mm.1 TC1100001570.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0