This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01305501_m1
See other LRIG2 GE Assays ›
Gene Symbol:
Lrig2
Gene Name:
leucine-rich repeats and immunoglobulin-like domains 2
Gene Aliases:
4632419I10Rik, BB096938
Chromosome Location:
Chr.3: 104453980 - 104511938 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01305501_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Lrig2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001025067.1 NP_001020238.1 13 - 14 1962 128
GenBank mRNA AK028527.1 - 3 - 4 579 128
AK036116.1 - 13 - 14 1964 128
AK173030.1 - 3 - 4 860 128
BC094228.1 - 13 - 14 1899 30362610 128
BC138422.1 - 13 - 14 1873 8861442 128

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0300002648.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Lrig2

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

sensory perception of sound
regulation of platelet-derived growth factor receptor signaling pathway
innervation
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0300020650.mm.1 NM_001025067.2 Refseq
Affymetrix Mouse Clariom S Array PSR0300020650.mm.1 TC0300002648.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0