This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01305483_m1
See other 9930111H07RIK GE Assays ›
Gene Symbol:
9930111H07Rik
Gene Name:
RIKEN cDNA 9930111H07 gene
Gene Aliases:
-
Chromosome Location:
Chr.1: 85775271 - 85784694 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01305483_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 9930111H07Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_108086.1 2 - 3 105 65
GenBank mRNA AK020686.1 - 2 - 3 106 65
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BY723014.1 - 2 - 3 106 Chr. 1 - 85775271 - 85784505 [+] on Build GRCm38 65

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream microRNA 8096 Mir8096 Gm27479,
mmu-mir-8096
-25.4kb Sense Chr. 1 - 85751948 - 85752074 on NCBI Build GRCm38
Downstream calcium binding protein 39 Cab39 AA408805,
AA960512,
C78372,
MO25,
MO25alpha
11.9kb Sense Chr. 1 - 85793410 - 85851577 on NCBI Build GRCm38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0100000769.mm.1

Set Membership:

Amplicon length less than or equal to 70 Mus musculus Probe spans exons

MicroArray Probes:

Agilent Affymetrix

Array Name Probe IDs Targets Target Type
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 21140 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 775430 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 204771 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04173217 - -
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P140364 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P04173217 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00949356 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20408423 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20408409 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00619437 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01110197 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20408415 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20408420 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20408412 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20408398 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20408400 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20408392 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20408421 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20408397 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20408411 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00666530 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20408413 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20408417 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20408403 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20408361 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20408394 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20408419 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20408407 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0100006411.mm.1 NR_108086.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100006411.mm.1 TC0100000769.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0