This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01305330_g1
See other F11R GE Assays ›
Gene Symbol:
F11r
Gene Name:
F11 receptor
Gene Aliases:
9130004G24, AA638916, ESTM33, JAM, JAM-1, JAM-A, Jcam, Jcam1, Ly106
Chromosome Location:
Chr.1: 171437561 - 171464593 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01305330_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: F11r
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_172647.2 NP_766235.1 4 - 5 464 79
GenBank mRNA AK033574.1 - 4 - 5 465 79
BC021876.1 - 4 - 5 407 3985305 79
CT010347.1 - 4 - 5 384 79

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0100001592.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene F11r

Gene Ontology Categories:

Function(s) Process(es)

cell adhesion
viral process
epithelial cell differentiation
actomyosin structure organization
negative regulation of GTPase activity
positive regulation of GTPase activity
positive regulation of blood pressure
intestinal absorption
establishment of protein localization to plasma membrane
establishment of endothelial intestinal barrier
regulation of membrane permeability
regulation of actin cytoskeleton reorganization
protein binding
PDZ domain binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0100013151.mm.1 NM_172647.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100013151.mm.1 NM_172647.2 Refseq
Affymetrix Mouse Clariom S Array PSR0100013151.mm.1 TC0100001592.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-9c87ff496-8j9ll:80/100.66.128.142:80.
git-commit: 146a63324c51a096ecccde4707f5fe4015d9ef66
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36_secrets_rm