This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01305304_m1
See other SPATS2 GE Assays ›
Gene Symbol:
Spats2
Gene Name:
spermatogenesis associated, serine-rich 2
Gene Aliases:
2700012F11Rik, 59kDa, Scr59, p59
Chromosome Location:
Chr.15: 99125926 - 99213215 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01305304_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Spats2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_139140.1 NP_631879.1 8 - 9 1149 107
GenBank mRNA AB079132.1 - 8 - 9 1149 107
AK048720.1 - 8 - 9 1191 107
AK159335.1 - 8 - 9 1244 107
AK167788.1 - 7 - 8 827 107
AK171266.1 - 7 - 8 905 107
AK215485.1 - 2 - 3 163 107
BC023432.1 - 6 - 7 799 5029821 107
BC072579.1 - 8 - 9 1416 107

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1500001008.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Spats2

Gene Ontology Categories:

Function(s) Process(es)

biological_process
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1500007838.mm.1 NM_139140.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500007838.mm.1 NM_139140.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500007838.mm.1 TC1500001008.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0