This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01305101_g1
See other DNASE1L2 GE Assays ›
Gene Symbol:
Dnase1l2
Gene Name:
deoxyribonuclease 1-like 2
Gene Aliases:
4733401H14Rik
Chromosome Location:
Chr.17: 24440087 - 24443765 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01305101_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Dnase1l2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_025718.3 NP_079994.2 1 - 2 97 75
GenBank mRNA AK003619.1 - 1 - 2 98 75
BC106188.1 - 1 - 2 61 30135288 75

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK003619.1, BC106188.1, NM_025718.3 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Dnase1l2

Gene Ontology Categories:

Function(s) Process(es)

DNA catabolic process, endonucleolytic
hair follicle development
corneocyte development
DNA metabolic process
DNA catabolic process
nucleic acid phosphodiester bond hydrolysis
nuclease activity
endonuclease activity
endodeoxyribonuclease activity
deoxyribonuclease activity
hydrolase activity
endodeoxyribonuclease activity, producing 5'-phosphomonoesters
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0