This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01304896_g1
See other ALDH6A1 GE Assays ›
Gene Symbol:
Aldh6a1
Gene Name:
aldehyde dehydrogenase family 6, subfamily A1
Gene Aliases:
1110038I05Rik, AI314632, Mmsdh
Chromosome Location:
Chr.12: 84430717 - 84450950 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01304896_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Aldh6a1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_134042.2 NP_598803.1 8 - 9 1054 106
GenBank mRNA AF297860.1 - 8 - 9 1054 106
AK033587.1 - 8 - 9 1081 106
AK082790.1 - 8 - 9 1108 106
AK128924.1 - 8 - 9 1086 106
AK147146.1 - 8 - 9 1108 106
AK155814.1 - 8 - 9 1105 106
AK169915.1 - 8 - 9 1094 106
AK170305.1 - 8 - 9 1107 106
AK171581.1 - 8 - 9 1108 106
AK171896.1 - 8 - 9 1097 106
AK184313.1 - 2 - 3 249 106
AK211862.1 - 2 - 3 249 106
AK215486.1 - 2 - 3 249 106
BC031148.1 - 8 - 9 1179 4975560 106
BC033440.1 - 7 - 8 1077 4212084 106

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1200002078.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Aldh6a1

Panther Classification:

Molecular Function -

dehydrogenase

Gene Ontology Categories:

Function(s) Process(es)

thymine catabolic process
valine metabolic process
valine catabolic process
metabolic process
beta-alanine catabolic process
thymine metabolic process
brown fat cell differentiation
oxidation-reduction process
aldehyde dehydrogenase (NAD) activity
methylmalonate-semialdehyde dehydrogenase (acylating) activity
oxidoreductase activity
oxidoreductase activity, acting on the aldehyde or oxo group of donors, NAD or NADP as acceptor
thiolester hydrolase activity
malonate-semialdehyde dehydrogenase (acetylating) activity
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1200014267.mm.1 NM_134042.3 Refseq
Affymetrix Mouse Clariom S Array PSR1200014267.mm.1 TC1200002078.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0