This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01304465_m1
See other KIF2A GE Assays ›
Gene Symbol:
Kif2a
Gene Name:
kinesin family member 2A
Gene Aliases:
C530030B14Rik, Kif2, Kns2, M-kinesin
Chromosome Location:
Chr.13: 106960585 - 107022228 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01304465_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Kif2a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001145779.1 NP_001139251.1 4 - 5 369 97
NM_008442.2 NP_032468.2 5 - 6 752 97
GenBank mRNA AK016720.1 - 4 - 5 372 97
AK037037.1 - 4 - 5 282 97
BC006803.1 - 4 - 5 364 3152883 97
D12644.1 - 5 - 6 753 97

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Kif2a

Panther Classification:

Molecular Function -

microtubule binding motor protein

Gene Ontology Categories:

Function(s) Process(es)

microtubule-based movement
microtubule depolymerization
cell cycle
mitotic spindle organization
mitotic nuclear division
multicellular organism development
nervous system development
cell differentiation
cell division
mitotic spindle assembly
nucleotide binding
microtubule motor activity
ATP binding
microtubule binding
ATPase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0