This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01304445_m1
See other MOCS2 GE Assays ›
Gene Symbol:
Mocs2
Gene Name:
molybdenum cofactor synthesis 2
Gene Aliases:
AI415403
Chromosome Location:
Chr.13: 114818237 - 114829420 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01304445_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mocs2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_013826.3 NP_038854.2 2 - 3 210 71
GenBank mRNA AK010112.1 - 2 - 3 190 71
BC024371.1 - 2 - 3 170 4983231 71

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK010112.1, BC024371.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Mocs2

Panther Classification:

Molecular Function -

transfer/carrier protein

Gene Ontology Categories:

Function(s) Process(es)

Mo-molybdopterin cofactor biosynthetic process
molybdopterin cofactor biosynthetic process
nucleotide binding
transferase activity
molybdopterin synthase activity
identical protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1300008274.mm.1 NM_001113374.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300008274.mm.1 NM_001113374.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300008274.mm.1 NM_001113375.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300008274.mm.1 NM_001113375.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300008274.mm.1 NM_013826.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300008274.mm.1 NM_013826.3 Refseq
Affymetrix Mouse Clariom S Array PSR1300008274.mm.1 TC1300001318.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0