This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01304417_g1
See other PGAP3 GE Assays ›
Gene Symbol:
Pgap3
Gene Name:
post-GPI attachment to proteins 3
Gene Aliases:
CAB2, D430035D22Rik, Perld1
Chromosome Location:
Chr.11: 98388672 - 98400539 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01304417_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pgap3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001033537.2 NP_001028709.2 7 - 8 991 78
GenBank mRNA AK052486.1 - 7 - 8 908 78
AK078974.1 - 7 - 8 917 78
AK170817.1 - 7 - 8 906 78
BC030368.1 - 5 - 6 611 78
BC035945.1 - 7 - 8 914 78

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100003674.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Pgap3

Gene Ontology Categories:

Function(s) Process(es)

GPI anchor metabolic process
GPI anchor biosynthetic process
hydrolase activity, acting on ester bonds

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28056218 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28056219 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28056220 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28056216 - -
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2018492 BC035945.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2018492 AK078974.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2018492 AK170817.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2018492 BC030368.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2018492 NM_001033537.2 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 412248 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 229055 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 583511 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 17800 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 883135 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 114151 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 445564 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P168282 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2018492 BC035945.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2018492 AK078974.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2018492 AK170817.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2018492 BC030368.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2018492 NM_001033537.2 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28056268 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28056271 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28056269 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28056257 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28056123 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28056264 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28056258 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28056265 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28056126 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28056122 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28056256 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28056121 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28056260 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28056266 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28056125 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28056261 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02406334 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P168282 - -
Whole Mouse Genome G4122A A_51_P491011 BC035945.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P491011 AK078974.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P491011 AK170817.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P491011 BC030368.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P491011 NM_001033537.2 mouse_transcriptome
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P168282 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28056218 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28056219 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28056216 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28056220 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28056168 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28056217 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28056083 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28056240 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28056152 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28056273 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28056268 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28056126 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28056075 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28056165 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28056141 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28056178 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28056265 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28056244 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28056116 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28056149 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28056190 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28056172 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28056119 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28056123 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28056191 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28056070 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28056251 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28056093 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28056184 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28056047 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28056235 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28056258 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28056224 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28056110 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28056199 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28056140 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28056219 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28056261 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28056078 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28056170 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28056272 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28056084 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28056210 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28056081 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28056157 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28056209 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28056222 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28056215 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28056253 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28056182 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28056193 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28056102 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28056046 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28056269 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28056107 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28056106 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28056096 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28056100 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28056167 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28056113 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28056186 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28056202 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28056125 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28056233 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28056264 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28056136 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28056213 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28056205 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28056134 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28056241 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28056206 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28056129 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28056143 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28056117 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28056238 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28056085 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28056161 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28056086 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28056089 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28056099 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28056266 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28056092 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28056180 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28056146 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28056242 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28056260 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28056079 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28056228 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28056072 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28056236 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28056088 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28056201 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28056127 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28056195 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28056207 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28056158 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28056076 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28056155 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28056174 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28056137 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28056139 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28056131 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28056197 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28056250 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28056194 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28056175 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28056121 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28056163 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28056122 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28056217 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28056171 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28056232 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28056201 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28056189 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09536861 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28056173 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28056161 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09499971 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09536823 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28056140 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28056180 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28056129 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09635492 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09396750 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09396752 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28056194 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28056158 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28056224 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09078013 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28056134 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28056149 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09675151 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28056121 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28056137 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28056239 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28056251 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28056222 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28056141 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09238424 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09437565 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28056174 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28056215 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28056146 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09536846 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09536843 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28056250 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09437558 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09536822 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28056221 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28056199 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28056242 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28056182 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28056241 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28056123 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28056184 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28056163 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28056152 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28056122 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09100050 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09499982 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28056202 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28056238 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28056157 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28056227 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09536834 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28056235 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28056178 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28056197 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09581310 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09536863 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28056143 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28056212 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28056209 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28056125 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28056216 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28056240 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09430401 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09238423 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28056230 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09702077 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28056226 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28056165 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10390626 - -
Affymetrix Mouse Gene 1.0 ST Array 10390625 - -
Affymetrix Mouse Gene 1.0 ST Array 10390634 - -
Affymetrix Mouse Gene 1.0 ST Array 10390623 - -
Affymetrix Mouse Gene 1.0 ST Array 10390624 - -
Affymetrix Mouse Gene 1.0 ST Array 10390632 - -
Affymetrix Mouse Gene 1.0 ST Array 10390633 - -
Affymetrix Mouse Gene 1.0 ST Array 10390629 - -
Affymetrix Mouse Gene 1.0 ST Array 10390627 - -
Affymetrix Mouse Gene 1.0 ST Array 10390628 - -
Affymetrix Mouse Gene 1.0 ST Array 10390630 - -
Affymetrix Mouse Gene 1.0 ST Array 10390631 - -
Affymetrix HT MG-430 PM Array Plate 1435037_PM_at - -
Affymetrix Mouse Gene 1.1 ST Array 10390622 - -
Mouse Genome 430 2.0 Array 1440613_PM_at - -
Mouse Genome 430 2.0 Array 1435037_PM_at - -
Mouse Genome 430 2.0 Array 1440613_at - -
Mouse Genome 430 2.0 Array 1435037_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 320655_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 320655_PM_at - -
Affymetrix Mouse Clariom S Array PSR1100033842.mm.1 NM_001033537.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100033842.mm.1 NM_001033537.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100033842.mm.1 TC1100003674.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100033843.mm.1 NM_001033537.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100033843.mm.1 NM_001033537.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100033843.mm.1 TC1100003674.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100033844.mm.1 NM_001033537.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100033844.mm.1 NM_001033537.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100033844.mm.1 TC1100003674.mm.1 MTA TC
Mouse Expression Set 430 1435037_at - -
Mouse Expression Set 430 1435037_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1435037_at - -
Affymetrix HT Mouse Genome 430B Array 1435037_PM_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 450585 - -
Array Name Probe IDs Targets Target Type
Illumina mouseRef-8 v1.1 expression beadchip ILMN_2711000 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2711000 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0