This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01303870_g1
See other DENND6B GE Assays ›
Gene Symbol:
Dennd6b
Gene Name:
DENN/MADD domain containing 6B
Gene Aliases:
1700027J05Rik, AI414881, Fam116b
Chromosome Location:
Chr.15: 89182213 - 89196474 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01303870_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Dennd6b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_027081.3 NP_081357.2 14 - 15 1280 64
GenBank mRNA AK006427.1 - 5 - 6 417 64
AK154949.1 - 11 - 12 1860 64
BC030395.1 - 14 - 15 1534 64
BC058958.1 - 10 - 11 787 64
BC080797.1 - 10 - 11 1169 64
BC139003.1 - 14 - 15 1254 8862025 64

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1500002042.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Dennd6b

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of GTPase activity
guanyl-nucleotide exchange factor activity
Rab guanyl-nucleotide exchange factor activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1500016348.mm.1 NM_027081.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500016348.mm.1 NM_027081.3 Refseq
Affymetrix Mouse Clariom S Array PSR1500016348.mm.1 TC1500002042.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0