This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01302895_g1
See other PGF GE Assays ›
Gene Symbol:
Pgf
Gene Name:
placental growth factor
Gene Aliases:
AI854365, PIGF, Plgf
Chromosome Location:
Chr.12: 85166637 - 85177785 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01302895_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pgf
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001271705.1 NP_001258634.1 1 - 2 88 90

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001271705.1 transcripts.

Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

growth factor

Gene Ontology Categories:

Function(s) Process(es)

angiogenesis
branching involved in ureteric bud morphogenesis
positive regulation of endothelial cell proliferation
sprouting angiogenesis
multicellular organism development
positive regulation of cell proliferation
cell differentiation
positive regulation of angiogenesis
positive regulation of cell division
regulation of morphogenesis of a branching structure
growth factor activity
protein homodimerization activity
protein heterodimerization activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1200014469.mm.1 NM_001271705.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200014469.mm.1 TC1200002089.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0