This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01302774_m1
See other DPF3 GE Assays ›
Gene Symbol:
Dpf3
Gene Name:
D4, zinc and double PHD fingers, family 3
Gene Aliases:
2810403B03Rik, 6530402L11Rik, BAF45C, C78788, CERD4, cer-d4
Chromosome Location:
Chr.12: 83213751 - 83487736 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01302774_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Dpf3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001267625.1 NP_001254554.1 7 - 8 770 89
NM_001267626.1 NP_001254555.1 7 - 8 770 89
NM_058212.2 NP_478119.1 7 - 8 767 89
GenBank mRNA AF362750.1 - 7 - 8 739 89
AK012970.1 - 7 - 8 775 89
AK039011.1 - 7 - 8 751 89
BC048572.1 - 7 - 8 650 6705809 89
FJ151218.1 - 7 - 8 742 89

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1200002045.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Dpf3

Panther Classification:

Molecular Function -

histone modifying enzyme

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of defense response to virus by host
transcription, DNA-templated
regulation of transcription, DNA-templated
nervous system development
chromatin modification
mitophagy in response to mitochondrial depolarization
zinc ion binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1200014047.mm.1 NM_001267625.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200014047.mm.1 NM_001267626.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200014047.mm.1 NM_058212.2 Refseq
Affymetrix Mouse Clariom S Array PSR1200014047.mm.1 TC1200002045.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1200014048.mm.1 NM_001267625.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200014048.mm.1 NM_001267626.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200014048.mm.1 NM_058212.2 Refseq
Affymetrix Mouse Clariom S Array PSR1200014048.mm.1 TC1200002045.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0