This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01302639_m1
See other PIGH GE Assays ›
Gene Symbol:
Pigh
Gene Name:
phosphatidylinositol glycan anchor biosynthesis, class H
Gene Aliases:
2210416H01Rik, A930028P05Rik
Chromosome Location:
Chr.12: 79080664 - 79089754 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01302639_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pigh
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_029988.2 NP_084264.1 3 - 4 525 93
GenBank mRNA AK167892.1 - 3 - 4 507 93
BC086806.1 - 3 - 4 476 30603877 93
BQ963876.1 - 1 - 2 84 93

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1200001976.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Pigh

Panther Classification:

Molecular Function -

glycosyltransferase

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function
transferase activity
transferase activity, transferring glycosyl groups
phosphatidylinositol N-acetylglucosaminyltransferase activity

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28567641 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28567644 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28567643 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28567640 - -
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P606826 BC086806.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P606826 AK030744.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P606826 AK167892.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P606826 NM_029988.2 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P606826 BC086806.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P606826 AK030744.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P606826 AK167892.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P606826 NM_029988.2 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 878833 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 262286 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 342692 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 309199 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 765846 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 765117 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 33488 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P109102 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P606826 BC086806.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P606826 AK030744.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P606826 AK167892.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P606826 NM_029988.2 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P606826 BC086806.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P606826 AK030744.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P606826 AK167892.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P606826 NM_029988.2 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28567641 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28567640 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28567643 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28567650 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28567659 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28567648 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28567634 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28567639 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28567655 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28567647 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28567658 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28567638 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28567661 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28567633 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28567654 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28567645 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28567631 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28567652 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P109102 - -
Whole Mouse Genome G4122A A_52_P606826 BC086806.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P606826 AK030744.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P606826 AK167892.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P606826 NM_029988.2 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P606826 BC086806.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P606826 AK030744.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P606826 AK167892.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P606826 NM_029988.2 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P260516 BC086806.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P260516 BY737882.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P260516 AK030744.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P260516 AK167892.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P260516 AK020902.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P129655 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P109102 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09978720 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09862932 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09862923 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09978718 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28567640 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28567643 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28567644 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28567641 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28567596 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28567553 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28567538 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28567612 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28567659 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28567639 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28567602 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28567613 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28567578 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28567582 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28567585 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28567551 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28567619 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28567534 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28567597 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28567532 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28567638 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28567515 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28567584 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28567562 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28567606 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28567580 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28567640 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28567629 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28567603 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28567668 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28567526 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28567610 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28567547 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28567645 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28567601 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28567607 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28567536 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28567545 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28567593 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28567623 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28567643 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28567670 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28567595 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28567556 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28567559 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28567666 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28567568 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28567633 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28567518 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28567528 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28567641 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28567522 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28567572 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28567542 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28567564 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28567627 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28567663 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28567550 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28567567 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28567575 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28567590 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28567652 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28567569 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28567566 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28567609 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28567650 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28567616 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28567529 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28567531 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28567648 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28567631 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28567599 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28567587 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28567524 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28567658 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28567614 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28567513 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28567552 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28567600 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28567622 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28567516 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28567563 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28567544 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28567647 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28567624 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28567623 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28567654 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P10230025 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P09913357 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28567566 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28567656 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P09913356 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P09889313 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28567625 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28567622 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28567562 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P10396913 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28567617 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P09978982 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28567578 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28567579 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28567670 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28567603 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P09862932 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P10230021 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28567626 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28567614 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28567609 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28567601 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P10314705 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28567561 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28567653 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28567638 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28567652 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P09930124 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28567639 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28567627 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28567658 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28567666 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28567648 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28567650 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28567647 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28567612 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28567665 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28567661 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28567645 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P10230008 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28567634 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28567629 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28567575 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28567668 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28567572 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28567636 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28567640 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28567663 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P10399413 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28567659 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28567610 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P10061908 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10401171 - -
Affymetrix Mouse Gene 1.0 ST Array 10401170 - -
Affymetrix Mouse Gene 1.0 ST Array 10401168 - -
Affymetrix Mouse Gene 1.0 ST Array 10401167 - -
Affymetrix Mouse Gene 1.0 ST Array 10401169 - -
Affymetrix HT MG-430 PM Array Plate 1430589_PM_at - -
Affymetrix HT MG-430 PM Array Plate 1455860_PM_at - -
Affymetrix Mouse Gene 1.1 ST Array 10401166 - -
Mouse Genome 430 2.0 Array 1455860_at - -
Mouse Genome 430 2.0 Array 1430589_PM_at - -
Mouse Genome 430 2.0 Array 1430589_at - -
Mouse Genome 430 2.0 Array 1455860_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 110417_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 110417_at - -
Affymetrix Mouse Clariom S Array PSR1200013555.mm.1 NM_029988.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200013555.mm.1 NM_029988.2 Refseq
Affymetrix Mouse Clariom S Array PSR1200013555.mm.1 TC1200001976.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1200013556.mm.1 NM_029988.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200013556.mm.1 NM_029988.2 Refseq
Affymetrix Mouse Clariom S Array PSR1200013556.mm.1 TC1200001976.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1200013554.mm.1 NM_029988.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200013554.mm.1 NM_029988.2 Refseq
Affymetrix Mouse Clariom S Array PSR1200013554.mm.1 TC1200001976.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1200013553.mm.1 NM_029988.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200013553.mm.1 NM_029988.2 Refseq
Affymetrix Mouse Clariom S Array PSR1200013553.mm.1 TC1200001976.mm.1 MTA TC
Mouse Expression Set 430 1455860_PM_at - -
Mouse Expression Set 430 1455860_at - -
Mouse Expression Set 430 1430589_PM_at - -
Mouse Expression Set 430 1430589_at - -
Mouse Genome 430A 2.0 Array 1455860_PM_at - -
Mouse Genome 430A 2.0 Array 1455860_at - -
Affymetrix HT Mouse Genome 430A Array 1455860_at - -
Affymetrix HT Mouse Genome 430A Array 1455860_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1430589_at - -
Affymetrix HT Mouse Genome 430B Array 1430589_PM_at - -
Array Name Probe IDs Targets Target Type
Illumina mouse-6 v1.0 expression beadchip 1500484 - -
Illumina mouseRef-8 v1.0 expression beadchip 0001500484 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_1232225 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_1232225 - -
mouse-6 v1.1 (Illumina) scl0110417.1_4-S - -
Illumina Sentrix mouse-6.v1 bead array 1500484 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl0110417.1_4-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 1500484 - -
MouseRef-8 v2 Expression BeadChip ILMN_1232225 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_1232225 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-655dcf6cc8-78hlp:80/100.66.128.142:80.
git-commit: 0d764ad91c70359c551b4d51a80efc6847a0ee54
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0