This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01302631_m1
See other C4BP GE Assays ›
Gene Symbol:
C4bp
Gene Name:
complement component 4 binding protein
Gene Aliases:
AI195242, C4bpa
Chromosome Location:
Chr.1: 130635921 - 130661618 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01302631_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: C4bp
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_007576.3 NP_031602.3 3 - 4 527 127
GenBank mRNA AB094485.1 - 508 127
AB094486.1 - 3 - 4 443 127
AK136735.1 - 3 - 4 441 127
AK149468.1 - 3 - 4 534 127
AK149477.1 - 3 - 4 444 127
AK162345.1 - 3 - 4 441 127
BC012257.1 - 3 - 4 499 4163203 127
M17122.1 - 3 - 4 541 127

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0100003041.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene C4bp

Gene Ontology Categories:

Function(s) Process(es)

immune system process
complement activation, classical pathway
innate immune response
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0100024384.mm.1 NM_007576.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100024384.mm.1 NM_007576.3 Refseq
Affymetrix Mouse Clariom S Array PSR0100024384.mm.1 TC0100003041.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100024383.mm.1 NM_007576.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100024383.mm.1 NM_007576.3 Refseq
Affymetrix Mouse Clariom S Array PSR0100024383.mm.1 TC0100003041.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0