This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01302148_m1
See other RHOC GE Assays ›
Gene Symbol:
Rhoc
Gene Name:
ras homolog gene family, member C
Gene Aliases:
AI324259, Arh9, Arhc
Chromosome Location:
Chr.3: 104789034 - 104794459 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01302148_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rhoc
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001291859.1 NP_001278788.1 1 - 2 66 74
GenBank mRNA BY315470.1 - 1 - 2 67 74

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001291859.1 transcripts.

Set Membership:

Amplicon length between 71 and 85 Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

small GTPase

Gene Ontology Categories:

Function(s) Process(es)

cytokinesis
signal transduction
small GTPase mediated signal transduction
positive regulation of I-kappaB kinase/NF-kappaB signaling
apical junction assembly
positive regulation of lipase activity
nucleotide binding
signal transducer activity
protein binding
GTP binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0