This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01301913_g1
See other ECEL1 GE Assays ›
Gene Symbol:
Ecel1
Gene Name:
endothelin converting enzyme-like 1
Gene Aliases:
DINE, xce
Chromosome Location:
Chr.1: 87142874 - 87157085 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01301913_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ecel1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001277925.1 NP_001264854.1 16 - 17 2368 73
NM_021306.3 NP_067281.2 16 - 17 2360 73
GenBank mRNA AB026294.1 - 15 - 16 2188 73
AK039467.1 - 16 - 17 2361 73
AK159365.1 - 16 - 17 2370 73
BC057569.1 - 15 - 16 2190 6413731 73

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0100002758.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ecel1

Panther Classification:

Molecular Function -

metalloprotease

Gene Ontology Categories:

Function(s) Process(es)

respiratory system process
proteolysis
neuropeptide signaling pathway
intracellular signal transduction
metalloendopeptidase activity
peptidase activity
metallopeptidase activity
hydrolase activity
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0100022259.mm.1 NM_001277925.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100022259.mm.1 NM_021306.3 Refseq
Affymetrix Mouse Clariom S Array PSR0100022259.mm.1 TC0100002758.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0