This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01300505_g1
See other DDX23 GE Assays ›
Gene Symbol:
Ddx23
Gene Name:
DEAD (Asp-Glu-Ala-Asp) box polypeptide 23
Gene Aliases:
3110082M05Rik, 4921506D17Rik
Chromosome Location:
Chr.15: 98645379 - 98662936 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01300505_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ddx23
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001080981.1 NP_001074450.1 14 - 15 2022 90

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1500002182.mm.1

Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Mus musculus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

RNA splicing, via transesterification reactions
RNA secondary structure unwinding
ATP-dependent RNA helicase activity
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1500017740.mm.1 NM_001080981.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500017740.mm.1 NM_001080981.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500017740.mm.1 TC1500002182.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1500017741.mm.1 NM_001080981.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500017741.mm.1 NM_001080981.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500017741.mm.1 TC1500002182.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0