This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01299818_m1
See other SLC4A8 GE Assays ›
Gene Symbol:
Slc4a8
Gene Name:
solute carrier family 4 (anion exchanger), member 8
Gene Aliases:
AW493845, C230026C11, NDCBE, kNBC-3
Chromosome Location:
Chr.15: 100761700 - 100823971 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01299818_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Slc4a8
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_021530.2 NP_067505.2 3 - 4 456 62
GenBank mRNA AF224508.1 - 3 - 4 402 62
AK034237.1 - 3 - 4 458 62
AK039404.1 - 3 - 4 421 62
AK171610.1 - 2 - 3 232 62
AK173014.1 - 3 - 4 290 62
BB626223.1 - 3 - 4 457 62
BC030388.1 - 3 - 4 418 5401143 62

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Slc4a8

Panther Classification:

Molecular Function -

secondary carrier transporter

Gene Ontology Categories:

Function(s) Process(es)

transport
ion transport
sodium ion transport
anion transport
embryo development
inorganic anion transport
bicarbonate transport
sodium ion transmembrane transport
regulation of intracellular pH
anion transmembrane transport
transporter activity
inorganic anion exchanger activity
anion transmembrane transporter activity
sodium:bicarbonate symporter activity
antiporter activity
anion:anion antiporter activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0