Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0100007241.mm.1 |
NM_134252.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100007241.mm.1 |
NM_134252.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100007241.mm.1 |
TC0100000846.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100007237.mm.1 |
NM_134252.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100007237.mm.1 |
NM_134252.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100007237.mm.1 |
TC0100000846.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100007238.mm.1 |
NM_134252.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100007238.mm.1 |
NM_134252.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100007238.mm.1 |
TC0100000846.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100007236.mm.1 |
NM_134252.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100007236.mm.1 |
NM_134252.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100007236.mm.1 |
TC0100000846.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100007242.mm.1 |
NM_134252.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100007242.mm.1 |
NM_134252.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100007242.mm.1 |
TC0100000846.mm.1 |
MTA TC |