This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01299443_m1
See other ARFGAP3 GE Assays ›
Gene Symbol:
Arfgap3
Gene Name:
ADP-ribosylation factor GTPase activating protein 3
Gene Aliases:
0610009H19Rik, 1810004P07Rik, 1810035F16Rik, 9130416J18Rik, Arfgap1
Chromosome Location:
Chr.15: 83299739 - 83350344 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01299443_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Arfgap3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_025445.4 NP_079721.2 4 - 5 490 141
GenBank mRNA AK007732.1 - 4 - 5 407 141
AK028990.1 - 4 - 5 491 141
AK075788.1 - 4 - 5 479 141
AK169107.1 - 4 - 5 400 141
BC004081.1 - 4 - 5 3592719 141
BC060369.1 - 4 - 5 417 6310619 141

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1500001971.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Arfgap3

Panther Classification:

Molecular Function -

GTPase-activating protein

Gene Ontology Categories:

Function(s) Process(es)

transport
protein transport
vesicle-mediated transport
positive regulation of GTPase activity
GTPase activator activity
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1500015554.mm.1 NM_025445.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500015554.mm.1 NM_025445.4 Refseq
Affymetrix Mouse Clariom S Array PSR1500015554.mm.1 TC1500001971.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0