This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01299369_m1
See other TRIM23 GE Assays ›
Gene Symbol:
Trim23
Gene Name:
tripartite motif-containing 23
Gene Aliases:
6330516O20Rik, AI450195, Arfd1
Chromosome Location:
Chr.13: 104178774 - 104203421 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01299369_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Trim23
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_030731.3 NP_109656.1 5 - 6 2942 103
GenBank mRNA AK029533.1 - 6 - 7 1091 103
AK030862.1 - 5 - 6 2943 103
AK036210.1 - 6 - 7 1091 103
AK039280.1 - 6 - 7 1102 103
AK042070.1 - 6 - 7 1093 103
AK088922.1 - 5 - 6 907 103
BC056390.1 - 6 - 7 1083 6854944 103
BC059017.1 - 6 - 7 1083 6848690 103

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1300001170.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Trim23

Panther Classification:

Molecular Function -

G-protein

Gene Ontology Categories:

Function(s) Process(es)

protein ADP-ribosylation
small GTPase mediated signal transduction
protein ubiquitination
nucleotide binding
GTPase activity
ubiquitin-protein transferase activity
GTP binding
zinc ion binding
UDP-N-acetylmuramoylalanyl-D-glutamyl-2,6-diaminopimelate-D-alanyl-D-alanine ligase activity
ligase activity
ribosomal S6-glutamic acid ligase activity
GDP binding
identical protein binding
coenzyme F420-0 gamma-glutamyl ligase activity
coenzyme F420-2 alpha-glutamyl ligase activity
metal ion binding
protein-glycine ligase activity
protein-glycine ligase activity, initiating
protein-glycine ligase activity, elongating
tubulin-glycine ligase activity
protein-glutamic acid ligase activity
tubulin-glutamic acid ligase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1300007413.mm.1 NM_030731.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300007413.mm.1 NM_030731.3 Refseq
Affymetrix Mouse Clariom S Array PSR1300007413.mm.1 TC1300001170.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0