This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01299274_m1
See other SLC26A8 GE Assays ›
Gene Symbol:
Slc26a8
Gene Name:
solute carrier family 26, member 8
Gene Aliases:
-
Chromosome Location:
Chr.17: 28637779 - 28691237 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01299274_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Slc26a8
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001290320.1 NP_001277249.1 5 - 6 906 64
GenBank mRNA BC027076.1 - 5 - 6 906 5361175 64

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1700001767.mm.1

Set Membership:

Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

transporter

Gene Ontology Categories:

Function(s) Process(es)

transport
ion transport
anion transport
chloride transport
multicellular organism development
spermatogenesis
sulfate transport
oxalate transport
cell differentiation
sperm motility
sperm capacitation
meiotic cell cycle
transmembrane transport
sulfate transmembrane transport
chloride transmembrane transport
organic acid transmembrane transport
chloride channel activity
secondary active sulfate transmembrane transporter activity
anion transmembrane transporter activity
sulfate transmembrane transporter activity
anion:anion antiporter activity
oxalate transmembrane transporter activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1700016312.mm.1 TC1700001767.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0