This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01299088_g1
See other MLC1 GE Assays ›
Gene Symbol:
Mlc1
Gene Name:
megalencephalic leukoencephalopathy with subcortical cysts 1 homolog (human)
Gene Aliases:
AW048630, BB074274, Kiaa0027-hp, LVM, MLC, VL, WKL1, mKIAA0027
Chromosome Location:
Chr.15: 88955884 - 88982687 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01299088_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mlc1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_133241.2 NP_573504.1 6 - 7 585 71
GenBank mRNA AF449425.1 - 6 - 7 1039 71
AK030396.1 - 6 - 7 584 71
AK043757.1 - 6 - 7 585 71
AK122190.1 - 6 - 7 1027 71
BC024719.1 - 6 - 7 585 5363238 71

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1500002033.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Ion Channel Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Mlc1

Gene Ontology Categories:

Function(s) Process(es)

transport
ion transport
protein transport
vesicle-mediated transport
positive regulation of intracellular transport
ion transmembrane transport
regulation of response to osmotic stress
protein oligomerization
cellular response to cholesterol
caveolin-mediated endocytosis
ion channel activity
protein binding
protein transporter activity
protein complex binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1500016128.mm.1 NM_133241.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500016128.mm.1 NM_133241.2 Refseq
Affymetrix Mouse Clariom S Array PSR1500016128.mm.1 TC1500002033.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56ffd697dc-88d4p:80/100.66.130.94:80.
git-commit: 082a6d415be982b1f92c35e55dd71609d4576d3e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0