This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01299048_m1
See other BAK1 GE Assays ›
Gene Symbol:
Bak1
Gene Name:
BCL2-antagonist/killer 1
Gene Aliases:
Bak, N-BAK1, N-Bak
Chromosome Location:
Chr.17: 27019812 - 27028839 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01299048_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Bak1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_007523.2 NP_031549.2 1 - 2 217 93
GenBank mRNA AK089220.1 - 1 - 2 155 93
BC057589.1 - 1 - 2 169 6411580 93
BY197389.1 - 1 - 2 218 93
CX564692.1 - 1 - 2 150 93
Y13231.1 - 1 - 2 218 93

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK089220.1, BC057589.1, NM_007523.2, Y13231.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Apoptosis Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Bak1

Gene Ontology Categories:

Function(s) Process(es)

leukocyte homeostasis
B cell homeostasis
B cell apoptotic process
release of cytochrome c from mitochondria
blood vessel remodeling
myeloid cell homeostasis
B cell negative selection
apoptotic process
activation of cysteine-type endopeptidase activity involved in apoptotic process
mitochondrial fusion
cell proliferation
negative regulation of cell proliferation
intrinsic apoptotic signaling pathway in response to DNA damage
activation of cysteine-type endopeptidase activity involved in apoptotic process by cytochrome c
response to fungus
response to mycotoxin
establishment or maintenance of transmembrane electrochemical gradient
response to gamma radiation
positive regulation of calcium ion transport into cytosol
negative regulation of gene expression
cytolysis
negative regulation of endoplasmic reticulum calcium ion concentration
negative regulation of peptidyl-serine phosphorylation
cellular response to UV
limb morphogenesis
regulation of apoptotic process
positive regulation of apoptotic process
regulation of protein homodimerization activity
regulation of protein heterodimerization activity
fibroblast apoptotic process
positive regulation of proteolysis
regulation of mitochondrial membrane permeability
post-embryonic camera-type eye morphogenesis
homeostasis of number of cells
regulation of cell cycle
regulation of mitochondrial membrane potential
vagina development
intrinsic apoptotic signaling pathway in response to endoplasmic reticulum stress
thymocyte apoptotic process
positive regulation of release of cytochrome c from mitochondria
apoptotic signaling pathway
extrinsic apoptotic signaling pathway in absence of ligand
activation of cysteine-type endopeptidase activity
positive regulation of endoplasmic reticulum unfolded protein response
apoptotic process involved in patterning of blood vessels
protein binding
heat shock protein binding
identical protein binding
protein homodimerization activity
metal ion binding
protein heterodimerization activity
chaperone binding
BH domain binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1700015855.mm.1 NM_007523.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700015855.mm.1 NM_007523.2 Refseq
Affymetrix Mouse Clariom S Array PSR1700015855.mm.1 TC1700001714.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1700015852.mm.1 NM_007523.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700015852.mm.1 NM_007523.2 Refseq
Affymetrix Mouse Clariom S Array PSR1700015852.mm.1 TC1700001714.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1700015853.mm.1 NM_007523.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700015853.mm.1 NM_007523.2 Refseq
Affymetrix Mouse Clariom S Array PSR1700015853.mm.1 TC1700001714.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 480967 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0