This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01298126_m1
See other FOXN1 GE Assays ›
Gene Symbol:
Foxn1
Gene Name:
forkhead box N1
Gene Aliases:
D11Bhm185e, Fkh19, HFH-11, Hfh11, Whn, nu, nude
Chromosome Location:
Chr.11: 78357577 - 78386922 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01298126_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Foxn1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001277290.1 NP_001264219.1 5 - 6 976 73
NM_008238.2 NP_032264.1 5 - 6 979 73
GenBank mRNA BC108980.1 - 4 - 5 864 40047616 73
BC108981.1 - 4 - 5 867 40047617 73
X81593.1 - 5 - 6 929 73

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100003295.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Foxn1

Gene Ontology Categories:

Function(s) Process(es)

hair follicle development
lymphocyte homeostasis
T cell lineage commitment
transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
multicellular organism development
cell proliferation
epidermis development
regulation of gene expression
hemopoiesis
cell differentiation
keratinocyte differentiation
regulation of T cell differentiation in thymus
nail development
T cell homeostasis
blood vessel morphogenesis
thymus development
epithelial cell proliferation
positive regulation of hair follicle development
lymphoid lineage cell migration into thymus
thymus epithelium morphogenesis
regulation of positive thymic T cell selection
RNA polymerase II transcription factor activity, sequence-specific DNA binding
DNA binding
transcription factor activity, sequence-specific DNA binding
sequence-specific DNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1100030369.mm.1 NM_001277290.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100030369.mm.1 NM_008238.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100030369.mm.1 TC1100003295.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0