This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01297252_g1
See other DHX29 GE Assays ›
Gene Symbol:
Dhx29
Gene Name:
DEAH (Asp-Glu-Ala-His) box polypeptide 29
Gene Aliases:
3732415M03, AU043276, Ddxx, E130202M19Rik
Chromosome Location:
Chr.13: 112927530 - 112969187 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01297252_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Dhx29
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_172594.2 NP_766182.2 9 - 10 1223 95
GenBank mRNA AK049530.1 - 9 - 10 1296 95
BC020331.1 - 9 - 10 1316 95
BC050229.1 - 9 - 10 1252 95
BC057112.1 - 9 - 10 1296 6417650 95
BC082319.1 - 9 - 10 1286 6417162 95

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1300001273.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Dhx29

Panther Classification:

Molecular Function -

RNA helicase

Gene Ontology Categories:

Function(s) Process(es)

RNA processing
translation
translational initiation
nucleotide binding
nucleic acid binding
translation initiation factor activity
ATP-dependent RNA helicase activity
helicase activity
ATP binding
ATP-dependent helicase activity
hydrolase activity
ribosomal small subunit binding
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1300008087.mm.1 NM_172594.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300008087.mm.1 NM_172594.2 Refseq
Affymetrix Mouse Clariom S Array PSR1300008087.mm.1 TC1300001273.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0