This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01297012_m1
See other SGSM3 GE Assays ›
Gene Symbol:
Sgsm3
Gene Name:
small G protein signaling modulator 3
Gene Aliases:
1810012I01Rik, AI428557, BB175482, Cip85, R75178, RUSC3, Rutbc3
Chromosome Location:
Chr.15: 80977765 - 81012290 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01297012_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Sgsm3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_134091.2 NP_598852.2 4 - 5 352 59
GenBank mRNA AK170806.1 - 4 - 5 319 59
AY382616.1 - 4 - 5 306 59
BC018197.1 - 4 - 5 312 4190407 59
BC024122.1 - 4 - 5 312 5097970 59
BC025561.1 - 4 - 5 290 5322980 59
BY242851.1 - 4 - 5 354 59

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Sgsm3

Panther Classification:

Molecular Function -

GTPase-activating protein

Gene Ontology Categories:

Function(s) Process(es)

cell cycle
cell cycle arrest
positive regulation of GTPase activity
positive regulation of protein catabolic process
plasma membrane to endosome transport
regulation of catalytic activity
GTPase activator activity
protein binding
Rab GTPase binding
GTPase regulator activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0