This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01297004_g1
See other SGSM3 GE Assays ›
Gene Symbol:
Sgsm3
Gene Name:
small G protein signaling modulator 3
Gene Aliases:
1810012I01Rik, AI428557, BB175482, Cip85, R75178, RUSC3, Rutbc3
Chromosome Location:
Chr.15: 80977765 - 81012290 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01297004_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Sgsm3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_134091.2 NP_598852.2 12 - 13 1563 59
GenBank mRNA AK142314.1 - 12 - 13 1531 59
AK170806.1 - 12 - 13 1530 59
AK183939.1 - 3 - 4 382 59
AY382616.1 - 12 - 13 1517 59
BC018197.1 - 12 - 13 1523 4190407 59
BC024122.1 - 12 - 13 1523 5097970 59
BC025561.1 - 12 - 13 1501 5322980 59

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1500000713.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Sgsm3

Panther Classification:

Molecular Function -

GTPase-activating protein

Gene Ontology Categories:

Function(s) Process(es)

cell cycle
cell cycle arrest
positive regulation of GTPase activity
positive regulation of protein catabolic process
plasma membrane to endosome transport
regulation of catalytic activity
GTPase activator activity
protein binding
Rab GTPase binding
GTPase regulator activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1500005433.mm.1 NM_134091.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500005433.mm.1 NM_134091.2 Refseq
Affymetrix Mouse Clariom S Array PSR1500005433.mm.1 TC1500000713.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1500005435.mm.1 NM_134091.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500005435.mm.1 NM_134091.2 Refseq
Affymetrix Mouse Clariom S Array PSR1500005435.mm.1 TC1500000713.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1500005436.mm.1 NM_134091.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500005436.mm.1 NM_134091.2 Refseq
Affymetrix Mouse Clariom S Array PSR1500005436.mm.1 TC1500000713.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1500005434.mm.1 NM_134091.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500005434.mm.1 NM_134091.2 Refseq
Affymetrix Mouse Clariom S Array PSR1500005434.mm.1 TC1500000713.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0