This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01297003_g1
See other SGSM3 GE Assays ›
Gene Symbol:
Sgsm3
Gene Name:
small G protein signaling modulator 3
Gene Aliases:
1810012I01Rik, AI428557, BB175482, Cip85, R75178, RUSC3, Rutbc3
Chromosome Location:
Chr.15: 80977765 - 81012290 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01297003_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Sgsm3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_134091.2 NP_598852.2 11 - 12 1435 74
GenBank mRNA AK142314.1 - 11 - 12 1403 74
AK170806.1 - 11 - 12 1402 74
AY382616.1 - 11 - 12 1389 74
BC018197.1 - 11 - 12 1395 4190407 74
BC024122.1 - 11 - 12 1395 5097970 74
BC025561.1 - 11 - 12 1373 5322980 74

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Sgsm3

Panther Classification:

Molecular Function -

GTPase-activating protein

Gene Ontology Categories:

Function(s) Process(es)

cell cycle
cell cycle arrest
positive regulation of GTPase activity
positive regulation of protein catabolic process
plasma membrane to endosome transport
regulation of catalytic activity
GTPase activator activity
protein binding
Rab GTPase binding
GTPase regulator activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0