This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01296537_g1
See other XYLT2 GE Assays ›
Gene Symbol:
Xylt2
Gene Name:
xylosyltransferase II
Gene Aliases:
E030002B02Rik, XT-II, XT2, xylT-II
Chromosome Location:
Chr.11: 94663845 - 94677518 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01296537_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Xylt2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_145828.3 NP_665827.2 3 - 4 893 84
GenBank mRNA AK083482.1 - 3 - 4 862 84
AK086804.1 - 3 - 4 825 84
AK150521.1 - 3 - 4 846 84
AK153332.1 - 3 - 4 846 84
AK158056.1 - 3 - 4 893 84
BC034082.1 - 2 - 3 412 4239646 84

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100003570.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Xylt2

Panther Classification:

Molecular Function -

transferase

Gene Ontology Categories:

Function(s) Process(es)

glycosaminoglycan biosynthetic process
proteoglycan biosynthetic process
heparin biosynthetic process
acetylglucosaminyltransferase activity
transferase activity
transferase activity, transferring glycosyl groups
protein xylosyltransferase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1100032712.mm.1 NM_145828.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100032712.mm.1 NM_145828.3 Refseq
Affymetrix Mouse Clariom S Array PSR1100032712.mm.1 TC1100003570.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0