This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01296454_m1
See other MYCBPAP GE Assays ›
Gene Symbol:
Mycbpap
Gene Name:
MYCBP associated protein
Gene Aliases:
4932408B01Rik, AMAP-1, AW125474
Chromosome Location:
Chr.11: 94501347 - 94532388 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01296454_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mycbpap
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_170671.2 NP_733771.2 1 - 2 103 106
GenBank mRNA BU840494.1 - 1 - 2 103 106

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100003565.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Mycbpap

Gene Ontology Categories:

Function(s)

multicellular organism development
spermatogenesis
biological_process
cell differentiation

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1100032632.mm.1 NM_170671.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100032632.mm.1 NM_170671.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100032632.mm.1 TC1100003565.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100032633.mm.1 NM_170671.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100032633.mm.1 NM_170671.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100032633.mm.1 TC1100003565.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0