This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01296370_m1
See other MYO1D GE Assays ›
Gene Symbol:
Myo1d
Gene Name:
myosin ID
Gene Aliases:
9930104H07Rik, AW544947, D11Ertd9e, myosin-1d
Chromosome Location:
Chr.11: 80482126 - 80780055 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01296370_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Myo1d
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_177390.3 NP_796364.2 19 - 20 2827 105
GenBank mRNA AK049374.1 - 19 - 20 2826 105
BC034071.1 - 5 - 6 807 105
BC039700.1 - 14 - 15 1959 105
BC150765.1 - 19 - 20 2693 9087676 105

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100003345.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Myo1d

Panther Classification:

Molecular Function -

actin binding motor protein

Gene Ontology Categories:

Function(s) Process(es)

metabolic process
negative regulation of phosphatase activity
cellular localization
microfilament motor activity
nucleotide binding
motor activity
actin binding
protein binding
calmodulin binding
ATP binding
protein domain specific binding
actin-dependent ATPase activity
calcium-dependent protein binding
actin filament binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1100030771.mm.1 NM_177390.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100030771.mm.1 NM_177390.3 Refseq
Affymetrix Mouse Clariom S Array PSR1100030771.mm.1 TC1100003345.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100030770.mm.1 TC1100003345.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0